Läs mer
N,N-Dimethylperfluoro-1-octanesulfonamide, TRC
Produktkod. 30375082
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30375082 100 mg
1 options
Produktkod. 30375082 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD486600100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N-Dimethylperfluorooctane Sulfonamide
CAS 213181-78-3
Molekylformel C10 H6 F17 N O2 S
Kvantitet 100 mg
Synonym N,N-Dimethylperfluoro-1-octanesulfonamide, N,N-Dimethylperfluorooctane Sulfonamide, N,N-Dimethylperfluoro-1-octane Sulfonamide, 1-Octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N,N-dimethyl- (9CI, ACI), 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N,N-dimethyl-1-octanesu
InChI formel InChI=1S/C10H6F17NO2S/c1-28(2)31(29,30)10(26,27)8(21,22)6(17,18)4(13,14)3(11,12)5(15,16)7(19,20)9(23,24)25/h1-2H3
LEDER CN(C)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N,N-dimethyloctane-1-sulfonamide
Molekylvikt (g/mol) 527.198
Formel vikt 526.985