Learn More
N,N-Dimethylpropionamide, ≥98%, Thermo Scientific Chemicals
Catalog Number p-7056097
Click to view available options
Quantity:
25 g
100 g
500 g
758-96-3
C5H11NO
101.149
MFCD00009301
MBHINSULENHCMF-UHFFFAOYSA-N
n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j
12965
N,N-dimethylpropanamide
CCC(=O)N(C)C
758-96-3
C5H11NO
101.149
MFCD00009301
MBHINSULENHCMF-UHFFFAOYSA-N
n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j
12965
N,N-dimethylpropanamide
CCC(=O)N(C)C
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.
Solubility
Soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -45°C (literature)
Color Colorless
Density 0.920 g/mL
Boiling Point 174°C to 175°C (literature)
Flash Point 62°C (144°F)
Linear Formula CH3CH2CON(CH3)2
Refractive Index 1.44
Quantity 25 g
Beilstein 1740719
Solubility Information Soluble in water.
Formula Weight 101.15
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material N,N-Dimethylpropionamide