Läs mer
N,N-Dinitroso-p-phenylenediamine-N,N-diacetic Acid, TRC
Produktkod. 30379936
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30379936 1 mg
1 options
Produktkod. 30379936 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4803751MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N-Dinitroso-p-phenylenediamine-N,N-diacetic Acid
CAS 6645-64-3
Molekylformel C10H10N4O6
Kvantitet 1 mg
InChI formel InChI=1S/C10H10N4O6.2Na/c15-9(16)5-13(11-19)7-1-2-8(4-3-7)14(12-20)6-10(17)18;;/h1-4H,5-6H2,(H,15,16)(H,17,18);;/q;2*+1/p-2
LEDER OC(=O)CN(N=O)c1ccc(cc1)N(CC(=O)O)N=O
Molekylvikt (g/mol) 282.21