Läs mer
N,N’-Diphenylsuberamide, TRC
Produktkod. 30372094
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30372094 500 mg
1 options
Produktkod. 30372094 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD491860500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-Diphenylsuberamide
CAS 14354-86-0
Molekylformel C20 H24 N2 O2
Kvantitet 500 mg
Synonym N1,N8-Diphenyloctanediamide
InChI formel InChI=1S/C20H24N2O2/c23-19(21-17-11-5-3-6-12-17)15-9-1-2-10-16-20(24)22-18-13-7-4-8-14-18/h3-8,11-14H,1-2,9-10,15-16H2,(H,21,23)(H,22,24)
LEDER O=C(CCCCCCC(=O)Nc1ccccc1)Nc2ccccc2
Molekylvikt (g/mol) 324.42
Formel vikt 324.1838