Läs mer
N,N’-(Ethylenedioxy)di-phthalimide, TRC
Produktkod. 30370038
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30370038 250 mg
1 options
Produktkod. 30370038 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE917765250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-(Ethylenedioxy)di-phthalimide
CAS 6437-67-8
Molekylformel C18 H12 N2 O6
Kvantitet 250 mg
Synonym 2,2'-[1,2-Ethanediylbis(oxy)]bis-1H-isoindole-1,3(2H)-dione
InChI formel InChI=1S/C18H12N2O6/c21-15-11-5-1-2-6-12(11)16(22)19(15)25-9-10-26-20-17(23)13-7-3-4-8-14(13)18(20)24/h1-8H,9-10H2
LEDER O=C1N(OCCON2C(=O)c3ccccc3C2=O)C(=O)c4ccccc14
Molekylvikt (g/mol) 352.3
Formel vikt 352.0695