Läs mer
N-(N-Methylpiperidinyl)methylindole, TRC
Produktkod. 30468024
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30468024 100 mg
1 options
Produktkod. 30468024 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM324860100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(N-Methylpiperidinyl)methylindole
CAS 137641-68-0
Molekylformel C15 H20 N2
Kvantitet 100 mg
Synonym 1H-Indole, 1-[(1-methyl-2-piperidinyl)methyl]-, 1-[(1-Methyl-2-piperidinyl)methyl]-1H-indole, 1-[(N-Methyl-2-piperidinyl)methyl]-1H-indole, N-(N-Methylpiperidinyl)methylindole, 1-(1-Methylpiperidin-2-yl)methylindole
InChI formel InChI=1S/C15H20N2/c1-16-10-5-4-7-14(16)12-17-11-9-13-6-2-3-8-15(13)17/h2-3,6,8-9,11,14H,4-5,7,10,12H2,1H3
LEDER CN1CCCCC1Cn2ccc3ccccc23
IUPAC-namn 1-[(1-methylpiperidin-2-yl)methyl]indole
Molekylvikt (g/mol) 228.33
Formel vikt 228.1626