Läs mer
N,N'-(N,N'-Dicarboxy-L-cystyl)di-glycine Dibenzyl N,N'-Di-tert-butyl Ester, TRC
Produktkod. 30374101
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30374101 2.5 mg
1 options
Produktkod. 30374101 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4309852.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-(N,N'-Dicarboxy-L-cystyl)di-glycine Dibenzyl N,N'-Di-tert-butyl Ester
CAS 32381-30-9
Molekylformel C34H46N4O10S2
Kvantitet 2.5 mg
Synonym N-[(1,1-dimethylethoxy)carbonyl]-L-cysteinyl-glycine Phenylmethyl Ester Bimol. (1-1')-disulfide
InChI formel InChI=1S/C34H46N4O10S2/c1-33(2,3)47-31(43)37-25(29(41)35-17-27(39)45-19-23-13-9-7-10-14-23)21-49-50-22-26(38-32(44)48-34(4,5)6)30(42)36-18-28(40)46-20-24-15-11-8-12-16-24/h7-16,25-26H,17-22H2,1-6H3,(H,35,41)(H,36,42)(H,37,43)(H,38,44)/t25-,26-/m0/s1
LEDER [C@@H](NC(OC(C)(C)C)=O)(C(NCC(OCC1=CC=CC=C1)=O)=O)CSSC[C@H](NC(OC(C)(C)C)=O)C(NCC(OCC2=CC=CC=C2)=O)=O
IUPAC-namn benzyl (6R,11R)-6-((2-(benzyloxy)-2-oxoethyl)carbamoyl)-11-((tert-butoxycarbonyl)amino)-2,2-dimethyl-4,12-dioxo-3-oxa-8,9-dithia-5,13-diazapentadecan-15-oate
Molekylvikt (g/mol) 734.88
Formel vikt 734.27