Läs mer
N,N’,N’’,N’’’-Tetraacetylchitotetraose, TRC
Produktkod. 30413690
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413690 1 mg
30411204 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30413690 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2765001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2706-65-2
Molekylformel C32 H54 N4 O21
Molekylvikt (g/mol) 830.79
Synonym N,N’,N’’,N’’’-Tetraacetyl Chitotetraose
IUPAC-namn N-[(2S,3R,4R,5S,6R)-2-[(2R,3S,4R,5R,6S)-5-acetamido-6-[(2R,3S,4R,5R,6S)-5-acetamido-6-[(2R,3S,4R,5R)-5-acetamido-1,2,4-trihydroxy-6-oxohexan-3-yl]oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
LEDER CC(=O)N[C@@H](C=O)[C@@H](O)[C@H](O[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3NC(=O)C)[C@H](O)[C@H]2NC(=O)C)[C@H](O)[C@H]1NC(=O)C)[C@H](O)CO
Specifikationer
Kemiskt namn eller material N,N',N' ',N' ''-Tetraacetylchitotetraose
Kvantitet 1 mg
InChI formel InChI=1S/C32H54N4O21/c1-10(42)33-14(5-37)22(47)27(15(46)6-38)55-31-20(35-12(3)44)25(50)29(17(8-40)53-31)57-32-21(36-13(4)45)26(51)28(18(9-41)54-32)56-30-19(34-11(2)43)24(49)23(48)16(7-39)52-30/h5,14-32,38-41,46-51H,6-9H2,1-4H3,(H,33,42)(H,34,43)(H,35,44)(H,36,45)/t14-,15+,16+,17+,18+,19+,20+,21+,22+,23+,24+,25+,26+,27+,28+,29+,30-,31-,32-/m0/s1
Formel vikt 830.3281