missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine, 99%, Thermo Scientific Chemicals

Catalog Number p-7058450
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
102-60-3
C14H32N2O4
292.42
MFCD00004534
NSOXQYCFHDMMGV-UHFFFAOYSA-N
quadrol, edetol, n,n,n',n'-tetrakis 2-hydroxypropyl ethylenediamine, entprol, adeka quadrol, quadrol l, neutrol te, 1,1',1,1'-ethylenedinitrilo tetra-2-propanol, edetolum latin, 2-propanol, 1,1',1,1'-1,2-ethanediyldinitrilo tetrakis
7615
1-[2-[bis(2-hydroxypropyl)amino]ethyl-(2-hydroxypropyl)amino]propan-2-ol
CC(CN(CCN(CC(C)O)CC(C)O)CC(C)O)O
This item is not returnable. View return policy
102-60-3
C14H32N2O4
292.42
MFCD00004534
NSOXQYCFHDMMGV-UHFFFAOYSA-N
quadrol, edetol, n,n,n',n'-tetrakis 2-hydroxypropyl ethylenediamine, entprol, adeka quadrol, quadrol l, neutrol te, 1,1',1,1'-ethylenedinitrilo tetra-2-propanol, edetolum latin, 2-propanol, 1,1',1,1'-1,2-ethanediyldinitrilo tetrakis
7615
1-[2-[bis(2-hydroxypropyl)amino]ethyl-(2-hydroxypropyl)amino]propan-2-ol
CC(CN(CCN(CC(C)O)CC(C)O)CC(C)O)O

N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine is used as a catalyst in the production of urethane foams. It acts as a complexing agent, plasticizer, surfactant solubilizer and curing agent for epoxy resin. It is also used as a viscosity modifier and as an anti-bloating agent for cattle. It is utilized in piezoelectric crystal detectors of sulfur dioxide and in additives for paint. It serves as a biological buffer as well as reagent for manganese. Further, it finds application in adhesives and sealant chemicals. In addition to this, it is used as a cross-linking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetrakis(2-hydroxypropyl)ethylenediamine is used as a catalyst in the production of urethane foams. It acts as a complexing agent, plasticizer, surfactant solubilizer and curing agent for epoxy resin. It is also used as a viscosity modifier and as an anti-bloating agent for cattle. It is utilized in piezoelectric crystal detectors of sulfur dioxide and in additives for paint. It serves as a biological buffer as well as reagent for manganese. Further, it finds application in adhesives and sealant chemicals. In addition to this, it is used as a cross-linking agent.

Solubility
Miscible with water, methanol, ethanol, toluene, ethylene glycol and perchloroethylene.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
TRUSTED_SUSTAINABILITY
Density 1.03
Boiling Point 240°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Refractive Index 1.48
Quantity 100 mL
Merck Index 14,3599
Solubility Information Miscible with water,methanol,ethanol,toluene,ethylene glycol and perchloroethylene.
Formula Weight 292.42
Percent Purity 99%
Chemical Name or Material N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'