Läs mer
N,N,N',N'-Tetramethyl-2-butene-1,4-diamine, TRC
Produktkod. 30425357
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425357 100 mg
30412765 500 mg
30425817 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425357 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT301100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4559-79-9
Molekylformel C8 H18 N2
Molekylvikt (g/mol) 142.24
Synonym N1,N1,N4,N4-Tetramethyl-2-butene-1,4-diamine, 1,4-Bis(dimethylamino)-2-butene
IUPAC-namn (E)-N,N,N',N'-tetramethylbut-2-ene-1,4-diamine
LEDER CN(C)C\C=C\CN(C)C
Specifikationer
Kemiskt namn eller material N,N,N',N'-Tetramethyl-2-butene-1,4-diamine
Kvantitet 100 mg
InChI formel InChI=1S/C8H18N2/c1-9(2)7-5-6-8-10(3)4/h5-6H,7-8H2,1-4H3/b6-5+
Formel vikt 142.147