Läs mer
N,N’,N’’-Triacetyl Chitotriose, TRC
Produktkod. 30450777
Change view
Kvantitet:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450777 50 mg
30453191 10 mg
30449661 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30450777 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT73500050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38864-21-0
Molekylformel C24H41N3O16
Molekylvikt (g/mol) 627.6
IUPAC-namn N-[(2S,3R,4R,5S,6R)-2-[(2R,3S,4R,5R,6S)-5-acetamido-6-[(2R,3S,4R,5R)-5-acetamido-1,2,4-trihydroxy-6-oxohexan-3-yl]oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
LEDER CC(=O)N[C@@H](C=O)[C@@H](O)[C@H](O[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2NC(=O)C)[C@H](O)[C@H]1NC(=O)C)[C@H](O)CO
Specifikationer
Kemiskt namn eller material N,N',N' '-Triacetylchitotriose
Kvantitet 50 mg
InChI formel InChI=1S/C24H41N3O16/c1-8(32)25-11(4-28)17(36)21(12(35)5-29)42-24-16(27-10(3)34)20(39)22(14(7-31)41-24)43-23-15(26-9(2)33)19(38)18(37)13(6-30)40-23/h4,11-24,29-31,35-39H,5-7H2,1-3H3,(H,25,32)(H,26,33)(H,27,34)/t11-,12+,13+,14+,15+,16+,17+,18+,19+,20+,21+,22+,23-,24-/m0/s1
Formel vikt 627.2487