missing translation for 'onlineSavingsMsg'
Läs mer

N,N,N-Trimethyl-3-[[(tridecafluorohexyl)sulfonyl]amino]-1-propanaminium, Inner Salt(>85%), TRC

Produktkod. 30456153
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456153 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30456153 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT90201025MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-[3-(perfluoro-1-hexanesulfonamido)propan-1-yl]-N,N,N-trimethylammonium
CAS 38850-51-0
Molekylformel C12 H15 F13 N2 O2 S
Kvantitet 25 mg
Synonym 1-Propanaminium','N,N,N-trimethyl-3-[[(tridecafluorohexyl)sulfonyl]amino]-','inner salt, N,N,N-Trimethyl-3-[[(tridecafluorohexyl)sulfonyl]amino]-1-propanaminium (N-TAmP-FHxSA), N-TAmP-FHxSA
InChI formel InChI=1S/C12H15F13N2O2S/c1-27(2,3)6-4-5-26-30(28,29)12(24,25)10(19,20)8(15,16)7(13,14)9(17,18)11(21,22)23/h4-6H2,1-3H3
LEDER C[N+](C)(C)CCC[N-]S(C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(=O)=O
IUPAC-namn 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexylsulfonyl-[3-(trimethylazaniumyl)propyl]azanide
Molekylvikt (g/mol) 498.3039
Formel vikt 498.0647
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.