Läs mer
N-(n-Nonyl)deoxynojirimycin, TRC
Produktkod. 30474886
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474886 10 mg
30476678 50 mg
30481775 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30474886 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN65030010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81117-35-3
Molekylformel C15 H31 N O4
Molekylvikt (g/mol) 289.41
Synonym N-Nonyldeoxynojirimycin
IUPAC-namn (2R,3R,4R,5S)-2-(hydroxymethyl)-1-nonylpiperidine-3,4,5-triol
LEDER CCCCCCCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO
Specifikationer
Kemiskt namn eller material N-(n-Nonyl)deoxynojirimycin
Kvantitet 10 mg
InChI formel InChI=1S/C15H31NO4/c1-2-3-4-5-6-7-8-9-16-10-13(18)15(20)14(19)12(16)11-17/h12-15,17-20H,2-11H2,1H3/t12-,13+,14-,15-/m1/s1
Formel vikt 289.2253