Learn More
N-(n-Propyl)maleimide, 94%, Thermo Scientific™
Catalog Number p-7043365
Click to view available options
Quantity:
5 g
25 g
21746-40-7
C7H9NO2
139.15
MFCD00014541
DABFKTHTXOELJF-UHFFFAOYSA-N
n-propylmaleimide, 1-propyl-1h-pyrrole-2,5-dione, n-n-propyl maleimide, n-propyl maleimide, n-n-propylmaleimide, 1h-pyrrole-2,5-dione, 1-propyl, 1-propyl-2,5-dihydro-1h-pyrrole-2,5-dione, n-propylmaleinimide, acmc-20ak14
89033
CCCN1C(=O)C=CC1=O
21746-40-7
C7H9NO2
139.15
MFCD00014541
DABFKTHTXOELJF-UHFFFAOYSA-N
n-propylmaleimide, 1-propyl-1h-pyrrole-2,5-dione, n-n-propyl maleimide, n-propyl maleimide, n-n-propylmaleimide, 1h-pyrrole-2,5-dione, 1-propyl, 1-propyl-2,5-dihydro-1h-pyrrole-2,5-dione, n-propylmaleinimide, acmc-20ak14
89033
CCCN1C(=O)C=CC1=O
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 24°C to 26°C
Density 1.073
Boiling Point 97°C (18 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.48
Quantity 5 g
UN Number UN2811
Beilstein 1525008
Solubility Information Insoluble in water.
IUPAC Name 1-propylpyrrole-2,5-dione
Formula Weight 139.15
Percent Purity 94%
Chemical Name or Material N-(n-Propyl)maleimide