Läs mer
N',N4-Bis(benzyloxycarbonyl)spermidine, TRC
Produktkod. 30336542
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30336542 5 mg
1 options
Produktkod. 30336542 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4109805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N',N4-Bis(benzyloxycarbonyl)spermidine
CAS 97628-87-0
Molekylformel C23H31N3O4
Kvantitet 5 mg
Synonym N-(4-Aminobutyl)-N-[3-[[(phenylmethoxy)carbonyl]amino]propyl]-carbamic Acid Phenylmethyl Ester, (4-Aminobutyl)[3-[[(phenylmethoxy)carbonyl]amino]propyl]-carbamic Acid Phenylmethyl Ester
InChI formel InChI=1S/C23H31N3O4/c24-14-7-8-16-26(23(28)30-19-21-12-5-2-6-13-21)17-9-15-25-22(27)29-18-20-10-3-1-4-11-20/h1-6,10-13H,7-9,14-19,24H2,(H,25,27)
LEDER NCCCCN(CCCNC(=O)OCc1ccccc1)C(=O)OCc2ccccc2
IUPAC-namn benzyl N-(4-aminobutyl)-N-[3-(phenylmethoxycarbonylamino)propyl]carbamate
Molekylvikt (g/mol) 413.51