Läs mer
N-Nitro-1,2,3,4-tetradehydro Argatroban Ethyl Ester, TRC
Produktkod. 17949534
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17949534 100 mg
17919914 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17949534 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN565120100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74874-09-2
Synonym ethyl (2R,4R)-4-methyl-1-[(2S)-2-[(3-methylquinolin-8-yl)sulfonylamino]-5-[(N-nitrocarbamimidoyl)amino]pentanoyl]piperidine-2-carboxylate
Specifikationer
Formatera Neat
Molekylformel C25H35N7O7S
MDL-nummer MFCD08458425
Molekylvikt (g/mol) 577.65, 577.66
InChI-nyckel DWOYDKNIEZWRDH-LSTHTHJFSA-N
LEDER CCOC(=O)[C@H]1C[C@H](C)CCN1C(=O)[C@H](CCC\N=C(/N)N[N+]([O-])=O)NS(=O)(=O)C1=CC=CC2=CC(C)=CN=C12
IUPAC-namn ethyl (2R,4R)-1-[(2S)-5-[(E)-[amino(nitroamino)methylidene]amino]-2-(3-methylquinoline-8-sulfonamido)pentanoyl]-4-methylpiperidine-2-carboxylate
Produkttyp Impurity