Läs mer
N-Nitro-Alpha-[2-(Methylamino)ethyl]benzyl Alcohol, TRC
Produktkod. 30179309
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179309 100 mg
30179261 250 mg
30178923 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179309 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN501015100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2659259-65-9
Molekylformel C10H14N2O2
Molekylvikt (g/mol) 194.23
Synonym N-(3-Hydroxy-3-phenylpropyl)-N-methylnitrous Amide
IUPAC-namn N-(3-hydroxy-3-phenylpropyl)-N-methylnitrous amide
LEDER CN(CCC(C1=CC=CC=C1)O)N=O
Specifikationer
Kemiskt namn eller material N-Nitro-alpha-[2-(Methylamino)ethyl]benzyl Alcohol
Kvantitet 100 mg
InChI formel InChI=1S/C10H14N2O2/c1-12(11-14)8-7-10(13)9-5-3-2-4-6-9/h2-6,10,13H,7-8H2,1H3
Formel vikt 194.11