Läs mer
N-Nitroso-3-pyrrolidinol, TRC
Produktkod. 30480316
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30480316 25 mg -
30480816 10 mg -
30478332 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480316 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN50101025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56222-35-6
Molekylformel C4H8N2O2
Molekylvikt (g/mol) 116.12
Synonym 1-Nitroso-3-pyrrolidinol, 3-Hydroxy-1-nitrosopyrrolidine, 3-Hydroxy-N-nitrosopyrrolidine, N-Nitroso-3-hydroxypyrrolidine
IUPAC-namn 1-nitrosopyrrolidin-3-ol
LEDER OC1CCN(C1)N=O
Specifikationer
Kemiskt namn eller material N-Nitroso-3-pyrrolidinol
Kvantitet 25 mg
InChI formel InChI=1S/C4H8N2O2/c7-4-1-2-6(3-4)5-8/h4,7H,1-3H2
Formel vikt 116.0586