Läs mer
N-Nitroso Acetanilide, TRC
Produktkod. 30483807
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30483807 100 mg
1 options
Produktkod. 30483807 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN501020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Nitroso Acetanilide
CAS 938-81-8
Molekylformel C8 H8 N2 O2
Kvantitet 100 mg
Synonym N-Nitroso-N-phenylacetamide, NSC 95272
InChI formel InChI=1S/C8H8N2O2/c1-7(11)10(9-12)8-5-3-2-4-6-8/h2-6H,1H3
LEDER CC(=O)N(N=O)c1ccccc1
IUPAC-namn N-nitroso-N-phenylacetamide
Molekylvikt (g/mol) 164.161
Formel vikt 164.059