Läs mer
N-Nitroso Desmethyl-chlorpromazine, TRC
Produktkod. 30428713
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30428713 100 mg
1 options
Produktkod. 30428713 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN703520100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Nitrosodesmethylchlorpromazine
CAS 55855-44-2
Molekylformel C16 H16 Cl N3 O S
Kvantitet 100 mg
Synonym 10H-Phenothiazine-10-propanamine','2-chloro-N-methyl-N-nitroso-, 2-Chloro-N-methyl-N-nitroso-10H-phenothiazine-10-propanamine, N-Nitrosodesmethylchlorpromazine, [3-(2-Chloro-phenothiazin-10-yl)-propyl]-methyl-nitroso-amine
InChI formel InChI=1S/C16H16ClN3OS/c1-19(18-21)9-4-10-20-13-5-2-3-6-15(13)22-16-8-7-12(17)11-14(16)20/h2-3,5-8,11H,4,9-10H2,1H3
LEDER CN(CCCN1c2ccccc2Sc3ccc(Cl)cc13)N=O
IUPAC-namn N-[3-(2-chlorophenothiazin-10-yl)propyl]-N-methylnitrous amide
Molekylvikt (g/mol) 333.836
Formel vikt 333.07