Läs mer
N-Nitroso Desmethyl-chlorpromazine, TRC
Produktkod. 30420822
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420822 50 mg
30418992 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30420822 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN70352050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Nitrosodesmethylchlorpromazine
CAS 55855-44-2
Molekylformel C16 H16 Cl N3 O S
Kvantitet 50 mg
Synonym 10H-Phenothiazine-10-propanamine','2-chloro-N-methyl-N-nitroso-, 2-Chloro-N-methyl-N-nitroso-10H-phenothiazine-10-propanamine, N-Nitrosodesmethylchlorpromazine, [3-(2-Chloro-phenothiazin-10-yl)-propyl]-methyl-nitroso-amine
InChI formel InChI=1S/C16H16ClN3OS/c1-19(18-21)9-4-10-20-13-5-2-3-6-15(13)22-16-8-7-12(17)11-14(16)20/h2-3,5-8,11H,4,9-10H2,1H3
LEDER CN(CCCN1c2ccccc2Sc3ccc(Cl)cc13)N=O
IUPAC-namn N-[3-(2-chlorophenothiazin-10-yl)propyl]-N-methylnitrous amide
Molekylvikt (g/mol) 333.836
Formel vikt 333.07