Läs mer
N-Nitroso-di-N-hexylamine, TRC
Produktkod. 30471814
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30471814 100 mg
1 options
Produktkod. 30471814 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN525575100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Nitrosodi-n-hexylamine
CAS 6949-28-6
Molekylformel C12 H26 N2 O
Kvantitet 100 mg
Synonym 1-Hexanamine','N-hexyl-N-nitroso- (9CI','ACI), Dihexylamine','N-nitroso- (7CI','8CI), N-Hexyl-N-nitroso-1-hexanamine (ACI), Di-n-Hexylnitrosamine, Dihexylnitrosamine, Dihexylnitrosoamine, N-Nitrosodi-n-hexylamine, N-Nitrosodihexylamine, NSC 18800, 1-Hexanamine','N-hexyl-N-nitroso- (9CI','ACI), Dihexylamine','N-nitroso- (7CI','8CI), N-Hexyl-N-nitroso-1-hexanamine (ACI), Di-n-Hexylnitrosamine, Dihexylnitrosamine, Dihexylnitrosoamine, N-Nitrosodi-n-hexylamine, N-Nitrosodihexylamine, NSC 18800
InChI formel InChI=1S/C12H26N2O/c1-3-5-7-9-11-14(13-15)12-10-8-6-4-2/h3-12H2,1-2H3
LEDER CCCCCCN(CCCCCC)N=O
IUPAC-namn N,N-dihexylnitrous amide
Molekylvikt (g/mol) 214.35
Formel vikt 214.2045