Läs mer
N-Nitroso Methyl Aniline Derivative, TRC
Produktkod. 30365001
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30365001 25 mg
1 options
Produktkod. 30365001 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC51145025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material [5-Chloro-2-(methylnitrosoamino)phenyl]phenylmethanone
CAS 51145-18-7
Molekylformel C14 H11 Cl N2 O2
Kvantitet 25 mg
Synonym [5-Chloro-2-(methylnitrosoamino)phenyl]phenylmethanone (ACI), Methanone, [5-chloro-2-(methylnitrosoamino)phenyl]phenyl- (9CI, ACI), N-(2-benzoyl-4-chlorophenyl)-N-methylnitrous amide
InChI formel InChI=1S/C14H11ClN2O2/c1-17(16-19)13-8-7-11(15)9-12(13)14(18)10-5-3-2-4-6-10/h2-9H,1H3
LEDER O=NN(C1=CC=C(Cl)C=C1C(=O)C=2C=CC=CC2)C
IUPAC-namn N-(2-benzoyl-4-chlorophenyl)-N-methylnitrous amide
Molekylvikt (g/mol) 274.7023
Formel vikt 274.0509