missing translation for 'onlineSavingsMsg'
Läs mer

N-Nitroso-N-ethyl Urea (Contains 40% water, 1.8 % AcOH), TRC

Produktkod. 30483948
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483948 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30483948 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN5261001G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material N-Nitroso-N-ethylurea
CAS 759-73-9
Anteckningar om renhetsgrad HPLC
Molekylformel C3 H7 N3 O2
Kvantitet 1 g
Synonym Urea','N-ethyl-N-nitroso- (9CI','ACI), N-Ethyl-N-nitrosourea (ACI), Urea','1-ethyl-1-nitroso- (7CI','8CI), 1-Ethyl-1-nitrosourea, ENU, N-Nitroso-N-ethylurea, NSC 45403
InChI formel InChI=1S/C3H7N3O2/c1-2-6(5-8)3(4)7/h2H2,1H3,(H2,4,7)
LEDER CCN(N=O)C(=O)N
IUPAC-namn 1-ethyl-1-nitrosourea
Molekylvikt (g/mol) 117.11
Formel vikt 117.0538
Procent renhet >95
Visa mer Visa mindre
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.