Läs mer
N-Nitroso-N-methyl-4-aminobutyric Acid Methyl Ester, TRC
Produktkod. 30465029
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30465029 100 mg
1 options
Produktkod. 30465029 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN529050100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Nitroso-N-methyl-4-aminobutyric Acid, Methyl Ester
CAS 51938-17-1
Molekylformel C6 H12 N2 O3
Kvantitet 100 mg
Synonym N-Nitroso-N-methyl-4-aminobutyric Acid Methyl Ester, Methyl 4-(methylnitrosoamino)butanoate, Butanoic acid, 4-(methylnitrosoamino)-, methyl ester, Methyl 4-(methylnitrosoamino)butanoate, Methyl(3-carboxypropyl)nitrosamine methyl ester
InChI formel InChI=1S/C6H12N2O3/c1-8(7-10)5-3-4-6(9)11-2/h3-5H2,1-2H3
LEDER COC(=O)CCCN(C)N=O
IUPAC-namn methyl 4-[methyl(nitroso)amino]butanoate
Molekylvikt (g/mol) 160.17
Formel vikt 160.0848