Läs mer
N-Nitroso-N-methyl-4-aminobutyric Acid, TRC
Produktkod. 17972795
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17972795 10 mg
17941705 25 mg
17921485 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17972795 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN52900010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61445-55-4
Molekylformel C5 H10 N2 O3
Molekylvikt (g/mol) 146.14
Synonym Butanoic acid, 4-(methylnitrosoamino)-, N-Nitroso-N-methyl-4-aminobutyric acid, NMBA, N-Nitrosomethylaminobutyric acid, 4-[Methyl(nitroso)amino]butanoic acid
IUPAC-namn 4-[methyl(nitroso)amino]butanoic acid
LEDER CN(CCCC(=O)O)N=O
Specifikationer
Kemiskt namn eller material N-Nitroso-N-methyl-4-aminobutyric Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C5H10N2O3/c1-7(6-10)4-2-3-5(8)9/h2-4H2,1H3,(H,8,9)
Formel vikt 146.0691
Procent renhet >95%