Läs mer
N-Nitroso-N-methylbenzylamine, TRC
Produktkod. 30468465
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30468465 500 mg
1 options
Produktkod. 30468465 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN529963500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Nitroso-N-methylbenzylamine
CAS 937-40-6
Anteckningar om renhetsgrad HPLC
Molekylformel C8 H10 N2 O
Kvantitet 500 mg
Synonym Benzenemethanamine, N-methyl-N-nitroso-, Benzylamine, N-methyl-N-nitroso- (6CI,7CI,8CI), N-Methyl-N-nitrosobenzenemethanamine, Benzyl(methyl)nitrosamine, Methyl(benzyl)nitrosamine, N-Methyl-N-benzylnitrosamine, N-Methyl-N-nitrosobenzylamine, N-Nitroso(benzyl)methylamine, N-Nitroso(methyl)benzylamine, N-Nitroso-N-methylbenzenemethanamine, N-Nitroso-N-methylbenzylamine, NMBA
InChI formel InChI=1S/C8H10N2O/c1-10(9-11)7-8-5-3-2-4-6-8/h2-6H,7H2,1H3
LEDER CN(Cc1ccccc1)N=O
IUPAC-namn N-benzyl-N-methylnitrous amide
Molekylvikt (g/mol) 150.1778
Formel vikt 150.0793