Läs mer
N-Nitroso-N-(phosphonomethyl)glycine, TRC
Produktkod. 30008710
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008710 100 mg
17933505 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008710 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN545150100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56516-72-4
Molekylformel C3 H7 N2 O6 P
Molekylvikt (g/mol) 198.07
Synonym Glyphosate-N-nitroso, Glycine, N-nitroso-N-(phosphonomethyl)- (9CI), 2-[Nitroso(phosphonomethyl)amino]acetic acid, N-Nitrosoglyphosate, Nitrosoglyphosate, Acetic acid, 2-[nitroso(phosphonomethyl)amino]-
IUPAC-namn 2-[nitroso(phosphonomethyl)amino]acetic acid
LEDER OC(=O)CN(CP(=O)(O)O)N=O
Specifikationer
Kemiskt namn eller material N-Nitroso-N-(phosphonomethyl)glycine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C3H7N2O6P/c6-3(7)1-5(4-8)2-12(9,10)11/h1-2H2,(H,6,7)(H2,9,10,11)
Kvantitet 100 mg
Formel vikt 198.0042
Procent renhet >95%