Läs mer
N-nitroso Rasagiline, TRC
Produktkod. 30134391
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134391 500 mg
30133482 250 mg
17970325 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134391 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN965525500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2470278-90-9
Molekylformel C12 H12 N2 O
Molekylvikt (g/mol) 200.237
Synonym N-Nitrosorasagiline, N-[(1R)-indan-1-yl]-N-prop-2-ynyl-nitrous amide, (1R)-N-nitroso-N-2-propyn-1-yl-2,3-dihydro-1H-inden-1-amine
IUPAC-namn N-[(1R)-2,3-dihydro-1H-inden-1-yl]-N-prop-2-ynylnitrous amide
LEDER O=NN(CC#C)[C@@H]1CCc2ccccc12
Specifikationer
Kemiskt namn eller material N-Nitrosorasagiline
Kvantitet 500 mg
InChI formel InChI=1S/C12H12N2O/c1-2-9-14(13-15)12-8-7-10-5-3-4-6-11(10)12/h1,3-6,12H,7-9H2/t12-/m1/s1
Formel vikt 200.095