Läs mer
N-Nitroso Tamsulosin Impurity, TRC
Produktkod. 30470530
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470530 10 mg
30470529 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30470530 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN39033010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Nitrosotamsulosin
CAS 2892260-31-8
Molekylformel C20 H27 N3 O6 S
Kvantitet 10 mg
Synonym (R)-5-(2-((2-(2-ethoxyphenoxy)ethyl)(nitroso)amino)propyl)-2-methoxybenzenesulfonamide, N-Nitroso Tamsulosin, N-Nitrosotamsulosin
InChI formel InChI=1S/C20H27N3O6S/c1-4-28-17-7-5-6-8-18(17)29-12-11-23(22-24)15(2)13-16-9-10-19(27-3)20(14-16)30(21,25)26/h5-10,14-15H,4,11-13H2,1-3H3,(H2,21,25,26)/t15-/m1/s1
LEDER CCOc1ccccc1OCCN(N=O)[C@H](C)Cc2ccc(OC)c(c2)S(=O)(=O)N
IUPAC-namn 5-[(2R)-2-[2-(2-ethoxyphenoxy)ethyl-nitrosoamino]propyl]-2-methoxybenzenesulfonamide
Molekylvikt (g/mol) 437.51
Formel vikt 437.162