missing translation for 'onlineSavingsMsg'
Läs mer

N-Nitrosoatenolol, Mikromol

Produktkod. 30457706
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30457706 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30457706 Leverantör Mikromol Leverantörsnummer MM0013.200025

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Nitrosoatenolol, Mikromol
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material N-Nitrosoatenolol
CAS 134720-04-0
Analyt- eller komponentnamn N-Nitrosoatenolol
Frakt skick Room Temperature
Molekylformel C14 H21 N3 O4
Certifikat och efterlevnad ISO 9001
Synonym Benzeneacetamide, 4-[2-hydroxy-3-[(1-methylethyl)nitrosoamino]propoxy]-, 4-[2-Hydroxy-3-[(1-methylethyl)nitrosoamino]propoxy]benzeneacetamide
InChI formel InChI=1S/C14H21N3O4/c1-10(2)17(16-20)8-12(18)9-21-13-5-3-11(4-6-13)7-14(15)19/h3-6,10,12,18H,7-9H2,1-2H3,(H2,15,19)
Lösningstyp Neat
LEDER CC(C)N(CC(O)COc1ccc(CC(=O)N)cc1)N=O
IUPAC-namn 2-[4-[2-hydroxy-3-[nitroso(propan-2-yl)amino]propoxy]phenyl]acetamide
Molekylvikt (g/mol) 295.33
Kvantitet 25 mg
Formel vikt 295.1532 g/mol
Visa mer Visa mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.