Läs mer
N-Nitrosodibenzylamine, TRC
Produktkod. 17902145
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17902145 5 mg
17952595 25 mg
17994995 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17902145 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN5254505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5336-53-8
Molekylformel C14 H14 N2 O
Molekylvikt (g/mol) 226.27
Synonym Dibenzylamine, N-nitroso- (6CI,7CI,8CI), N-Nitroso-N-(phenylmethyl)benzenemethanamine, Dibenzylnitrosamine, N,N-Dibenzylnitrosamine, N-Nitrosodibenzylamine, NSC 338, Nitrosodibenzylamine
IUPAC-namn N,N-dibenzylnitrous amide
LEDER O=NN(Cc1ccccc1)Cc2ccccc2
Specifikationer
Kemiskt namn eller material N-Nitrosodibenzylamine
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C14H14N2O/c17-15-16(11-13-7-3-1-4-8-13)12-14-9-5-2-6-10-14/h1-10H,11-12H2
Formel vikt 226.1106
Procent renhet >95%