Läs mer
N-Nitrosodimethylamine 0.2 mg/ml in Methanol, Mikromol
Produktkod. 30458682
Change view
Kvantitet:
1 ml
Produktkod. Kvantitet
30458682 1 ml
1 options
Produktkod. 30458682 Leverantör Mikromol Leverantörsnummer MM0487.09AME00.20
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Nitrosodimethylamine 0.2 mg/ml in Methanol, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Nitrosodimethylamine
CAS 62-75-9
Analyt- eller komponentnamn N-Nitrosodimethylamine
Frakt skick Ice pack (-20°C)
Molekylformel C2 H6 N2 O
Koncentration 0.2 mg/mL
Certifikat och efterlevnad ISO 17034
Synonym Methanamine, N-methyl-N-nitroso-, Dimethylamine, N-nitroso- (8CI), N-Methyl-N-nitrosomethanamine, DMN, DMNA, Dimethylnitrosamine, N-Nitroso-N,N-dimethylamine, N-Nitrosodimethylamine, NDMA, NSC 23226, N-Nitroso-dimethylamine
InChI formel InChI=1S/C2H6N2O/c1-4(2)3-5/h1-2H3
Lösningstyp Single Solution
LEDER CN(C)N=O
IUPAC-namn N,N-dimethylnitrous amide
Lösningsmedel eller matris Methanol
Molekylvikt (g/mol) 74.08
Kvantitet 1 ml
Formel vikt 74.048 g/mol