Läs mer
N-Nitrosodimethylamine (5 mg/mL in Methanol), TRC
Specifications
Specifications
| Kemiskt namn eller material | N-Nitrosodimethylamine |
| CAS | 62-75-9 |
| Molekylformel | C2 H6 N2 O |
| Kvantitet | 1x1 mL |
| Synonym | Methanamine, N-methyl-N-nitroso-, Dimethylamine, N-nitroso- (8CI), N-Methyl-N-nitrosomethanamine, DMN, DMNA, Dimethylnitrosamine, N-Nitroso-N,N-dimethylamine, N-Nitrosodimethylamine, NDMA, NSC 23226, N-Nitroso-dimethylamine |
| InChI formel | InChI=1S/C2H6N2O/c1-4(2)3-5/h1-2H3 |
| LEDER | CN(C)N=O |
| IUPAC-namn | N,N-dimethylnitrous amide |
| Molekylvikt (g/mol) | 74.08 |
| Formel vikt | 74.048 |
| Show More |