Läs mer
N-Nitrosoiminodibenzyl, TRC
Produktkod. 30476108
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476108 100 mg
30465878 50 mg
30467038 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476108 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN137450100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7458-08-4
Molekylformel C14H12N2O
Molekylvikt (g/mol) 224.26
Synonym 10,11-Dihydro-5-nitroso-5H-dibenz[b,f]azepine
IUPAC-namn 11-nitroso-5,6-dihydrobenzo[b][1]benzazepine
LEDER O=NN1c2ccccc2CCc3ccccc13
Specifikationer
Kemiskt namn eller material N-Nitrosoiminodibenzyl
Kvantitet 100 mg
InChI formel InChI=1S/C14H12N2O/c17-15-16-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)16/h1-8H,9-10H2
Formel vikt 224.095