Läs mer
N-Nitrosopiperazine, TRC
Produktkod. 17954925
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17954925 50 mg
17960685 500 mg
17963815 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17954925 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN54535050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5632-47-3
Molekylformel C4 H9 N3 O
Molekylvikt (g/mol) 115.13
Synonym Piperazine, 1-nitroso-, 1-Nitrosopiperazine, N-Mononitrosopiperazine, N-Nitrosopiperazine, NSC 50269, NSC 525340
IUPAC-namn 1-nitrosopiperazine
LEDER O=NN1CCNCC1
Specifikationer
Kemiskt namn eller material N-Nitrosopiperazine
Kvantitet 50 mg
InChI formel InChI=1S/C4H9N3O/c8-6-7-3-1-5-2-4-7/h5H,1-4H2
Formel vikt 115.0746