Läs mer
N-Nitrososarcosine, TRC
Produktkod. 30041157
Change view
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30041157 100 mg
30010549 1 g
18052140 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30041157 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN546650100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13256-22-9
Molekylformel C3 H6 N2 O3
Molekylvikt (g/mol) 118.09
Synonym Acetic acid, 2-(methylnitrosoamino)-, Glycine, N-methyl-N-nitroso- (9CI), Sarcosine, N-nitroso- (6CI,7CI,8CI), 2-(Methylnitrosoamino)acetic acid, N-Methyl-N-(carboxymethyl)nitrosamine, N-Nitrososarcosine
IUPAC-namn 2-[methyl(nitroso)amino]acetic acid
LEDER CN(CC(=O)O)N=O
Specifikationer
Kemiskt namn eller material N-Nitrososarcosine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C3H6N2O3/c1-5(4-8)2-3(6)7/h2H2,1H3,(H,6,7)
Formel vikt 118.0378
Procent renhet >95%