Läs mer
N,O-Bis(benzyloxycarbonyl) Halofuginone, TRC
Produktkod. 30339952
Change view
Kvantitet:
2 mg
Produktkod. Kvantitet
30339952 2 mg
1 options
Produktkod. 30339952 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4108952MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,O-Bis(benzyloxycarbonyl) Halofuginone
CAS 1246816-41-0
Molekylformel C32H29BrClN3O7
Kvantitet 2 mg
Synonym (2R,3S)-rel-2-[3-(7-Bromo-6-chloro-4-oxo-3(4H)-quinazolinyl)-2-oxopropyl]-3-[[(phenylmethoxy)carbonyl]oxy]-1-piperidinecarboxylic Acid Phenylmethyl Ester
InChI formel InChI=1S/C32H29BrClN3O7/c33-25-16-27-24(15-26(25)34)30(39)36(20-35-27)17-23(38)14-28-29(44-32(41)43-19-22-10-5-2-6-11-22)12-7-13-37(28)31(40)42-18-21-8-3-1-4-9-21/h1-6,8-11,15-16,20,28-29H,7,12-14,17-19H2/t28-,29+/m0/s1
LEDER Clc1cc2C(=O)N(CC(=O)C[C@H]3[C@@H](CCCN3C(=O)OCc4ccccc4)OC(=O)OCc5ccccc5)C=Nc2cc1Br
IUPAC-namn benzyl (2S,3R)-2-[3-(7-bromo-6-chloro-4-oxoquinazolin-3-yl)-2-oxopropyl]-3-phenylmethoxycarbonyloxypiperidine-1-carboxylate
Molekylvikt (g/mol) 682.95
Formel vikt 681.0877