Läs mer
N,O-Ditrityl Losartan, TRC
Produktkod. 30370681
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30370681 25 mg
30373511 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30370681 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD49432025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1796930-34-1
Molekylformel C60 H51 Cl N6 O
Molekylvikt (g/mol) 907.54
Synonym 2H-Tetrazole, 5-[4'-[[2-butyl-4-chloro-5-[(triphenylmethoxy)methyl]-1H-imidazol-1-yl]methyl][1,1'-biphenyl]-2-yl]-2-(triphenylmethyl)-, 5-[4'-[[2-Butyl-4-chloro-5-[[(triphenylmethyl)oxy]methyl]-1H-imidazol-1-yl]methyl]biphenyl-2-yl]-2-(triphenylmethyl)tetrazole, 5-[4'-[[2-Butyl-4-chloro-5-[(triphenylmethoxy)methyl]-1H-imidazol-1-yl]methyl][1,1'-biphenyl]-2-yl]-2-(triphenylmethyl)-2H-tetrazole, 2H-Tetrazole, 5-[4'-[[2-butyl-4-chloro-5-[(triphenylmethoxy)methyl]-1H-imidazol-1-yl]methyl][1,1'-biphenyl]-2-yl]-2-(triphenylmethyl)-, 5-[4'-[[2-Butyl-4-chloro-5-[[(triphenylmethyl)oxy]methyl]-1H-imidazol-1-yl]methyl]biphenyl-2-yl]-2-(triphenylmethyl)tetrazole
IUPAC-namn 5-[2-[4-[[2-butyl-4-chloro-5-(trityloxymethyl)imidazol-1-yl]methyl]phenyl]phenyl]-2-trityltetrazole
LEDER CCCCc1nc(Cl)c(COC(c2ccccc2)(c3ccccc3)c4ccccc4)n1Cc5ccc(cc5)c6ccccc6c7nnn(n7)C(c8ccccc8)(c9ccccc9)c%10ccccc%10
Specifikationer
Kemiskt namn eller material N,O-Ditrityl Losartan
Kvantitet 25 mg
InChI formel InChI=1S/C60H51ClN6O/c1-2-3-38-56-62-57(61)55(44-68-60(50-30-16-7-17-31-50,51-32-18-8-19-33-51)52-34-20-9-21-35-52)66(56)43-45-39-41-46(42-40-45)53-36-22-23-37-54(53)58-63-65-67(64-58)59(47-24-10-4-11-25-47,48-26-12-5-13-27-48)49-28-14-6-15-29-49/h4-37,39-42H,2-3,38,43-44H2,1H3
Formel vikt 906.3813