missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Octane, Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Melting Point | -57°C |
Color | Colorless |
Boiling Point | 125°C |
Packaging | Amber glass bottle |
Quantity | 100 mL |
Vapor Density | 3.9 |
Formula Weight | 114.23g/mol |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'