Läs mer
N-Octanoylglycine, TRC
Produktkod. 30414272
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30414272 10 g
1 options
Produktkod. 30414272 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO23973510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Octanoylglycine
CAS 14246-53-8
Molekylformel C10 H19 N O3
Kvantitet 10 g
Synonym Glycine','N-(1-oxooctyl)- (9CI','ACI), Glycine','N-octanoyl- (6CI','7CI','8CI), N-(1-Oxooctyl)glycine (ACI), 2-Octanamidoacetic acid, Capryloylglycine, Lipacide C 8G, N-Octanoyl-glycine, Octanoylglycine, N-Octanoylglycine
InChI formel InChI=1S/C10H19NO3/c1-2-3-4-5-6-7-9(12)11-8-10(13)14/h2-8H2,1H3,(H,11,12)(H,13,14)
LEDER CCCCCCCC(=O)NCC(=O)O
IUPAC-namn 2-(octanoylamino)acetic acid
Molekylvikt (g/mol) 201.26
Formel vikt 201.1365