missing translation for 'onlineSavingsMsg'
Läs mer

n-Octyl Chloroformate, TRC

Produktkod. 30416471
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30416471 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30416471 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO23998010G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material n-Octyl Chloroformate
CAS 7452-59-7
Molekylformel C9H17ClO2
Kvantitet 10 g
Synonym Chloroformic Acid Octyl Ester, 1-OctylChloroformate, Octyl Chloroformate, n-Octyl Chloroformate
InChI formel InChI=1S/C9H17ClO2/c1-2-3-4-5-6-7-8-12-9(10)11/h2-8H2,1H3
LEDER CCCCCCCCOC(=O)Cl
IUPAC-namn octyl carbonochloridate
Molekylvikt (g/mol) 192.68
Formel vikt 192.0917
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.