Läs mer
N-[p-(Acetylmercuric)phenyl]maleamate, TRC
Produktkod. 30326885
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30326885 250 mg
1 options
Produktkod. 30326885 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA186290250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[p-(Acetylmercuric)phenyl]maleamate
CAS 134998-13-3
Molekylformel C12H11HgNO5
Kvantitet 250 mg
Synonym (Z)-4-Oxo-4-(phenylamino)-2-butenoic Acid Mercury Complex, (Z)-(Acetato-O)[4-[(3-carboxy-1-oxo-2-propenyl)amino]phenyl]mercury
InChI formel InChI=1S/C10H8NO3.C2H4O2.Hg/c12-9(6-7-10(13)14)11-8-4-2-1-3-5-8;1-2(3)4;/h2-7H,(H,11,12)(H,13,14);1H3,(H,3,4);/q;;+1/p-1/b7-6-;;
LEDER O=C(/C=C\C(O)=O)NC1=CC=C([Hg]OC(C)=O)C=C1
IUPAC-namn acetyloxy-[4-[[(Z)-3-carboxyprop-2-enoyl]amino]phenyl]mercury
Molekylvikt (g/mol) 449.81