Läs mer
N-[p-(Acetylmercuric)phenyl]maleimide, TRC
Produktkod. 30320316
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30320316 25 mg
1 options
Produktkod. 30320316 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18628025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[p-(Acetylmercuric)phenyl]maleimide
CAS 117259-61-7
Molekylformel C12H9HgNO4
Kvantitet 25 mg
Synonym (Acetato-O)[4-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)phenyl]mercury, 1-Phenyl-1H-Pyrrole-2,5-dione Mercury Complex
InChI formel InChI=1S/C10H6NO2.C2H4O2.Hg/c12-9-6-7-10(13)11(9)8-4-2-1-3-5-8;1-2(3)4;/h2-7H;1H3,(H,3,4);/q;;+1/p-1
LEDER CC(=O)O[Hg]c1ccc(cc1)N2C(=O)C=CC2=O
IUPAC-namn acetyloxy-[4-(2,5-dioxopyrrol-1-yl)phenyl]mercury
Molekylvikt (g/mol) 431.79
Formel vikt 433.0238