Läs mer
N-p-Coumaroyltyrosine (~98%), TRC
Produktkod. 30366316
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366316 10 mg
30365852 50 mg
30349674 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366316 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC89998010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 77201-66-2
Molekylformel C18H17NO5
Molekylvikt (g/mol) 327.33
Synonym N-[(2E)-3-(4-Hydroxyphenyl)-1-oxo-2-propen-1-yl]-L-tyrosine, N-[(2E)-3-(4-hydroxyphenyl)-1-oxo-2-propenyl]-L-tyrosine, (E)-N-[3-(4-Hydroxyphenyl)-1-oxo-2-propenyl]-L-tyrosine
IUPAC-namn (2S)-3-(4-hydroxyphenyl)-2-[[(E)-3-(4-hydroxyphenyl)prop-2-enoyl]amino]propanoic acid
LEDER OC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)\C=C\c2ccc(O)cc2
Specifikationer
Kemiskt namn eller material N-p-Coumaroyltyrosine
Kvantitet 10 mg
InChI formel InChI=1S/C18H17NO5/c20-14-6-1-12(2-7-14)5-10-17(22)19-16(18(23)24)11-13-3-8-15(21)9-4-13/h1-10,16,20-21H,11H2,(H,19,22)(H,23,24)/b10-5+/t16-/m0/s1
Formel vikt 327.1107