Läs mer
N-fenyl-n'-cyklohexyl-p-fenylendiamin (>90 %), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Cyclohexyl-N'-phenyl-p-phenylenediamine (CPPD) |
| CAS | 101-87-1 |
| Molekylformel | C18 H22 N2 |
| Kvantitet | 500 mg |
| Synonym | 1,4-Benzenediamine, N-cyclohexyl-N'-phenyl- (9CI), p-Phenylenediamine, N-cyclohexyl-N'-phenyl- (6CI,8CI), N1-Cyclohexyl-N4-phenyl-1,4-benzenediamine, 1-N-Cyclohexyl-4-N-phenylbenzene-1,4-diamine, 4-(Cyclohexylamino)diphenylamine, 4020NA, ASM 4010, Antioxidant 4010, Antioxidant CPPD, Diafen FTs, Flexzone 6H, N-Cyclohexyl-N'-phenyl-1,4-benzenediamine, N-Cyclohexyl-N'-phenyl-1,4-phenylenediamine, N-Cyclohexyl-N'-phenyl-p-phenylenediamine, N-Phenyl-N'-cyclohexyl-p-phenylenediamine, N-Phenyl-N'-cyclohexylparaphenylenediamine, NA 4020, Nocrac 810, Santoflex CP, Vanax 6H, N-Cyclohexyl-N'-phenyl-p-phenylenediamine (CPPD) |
| InChI formel | InChI=1S/C18H22N2/c1-3-7-15(8-4-1)19-17-11-13-18(14-12-17)20-16-9-5-2-6-10-16/h1,3-4,7-8,11-14,16,19-20H,2,5-6,9-10H2 |
| LEDER | C1CCC(CC1)Nc2ccc(Nc3ccccc3)cc2 |
| IUPAC-namn | 4-N-cyklohexyl-1-N-fenylbensen-1,4-diamin |
| Molekylvikt (g/mol) | 266.3807 |
| Formel vikt | 266.1783 |