Läs mer
N-fenyl-p-fenylendiamin (p-anilinoanilin), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Phenyl-1,4-phenylenediamine |
| CAS | 101-54-2 |
| Molekylformel | C12 H12 N2 |
| Kvantitet | 25 g |
| Synonym | 1,4-Benzenediamine, N-phenyl- (9CI), Variamine Blue RT (6CI), p-Phenylenediamine, N-phenyl- (8CI), N1-Phenyl-1,4-benzenediamine, (4-(Phenylamino)phenyl)amine, 4-(N-Phenylamino)aniline, 4-(Phenylamino)aniline, 4-Amino-N-phenylaniline, 4-Aminodiphenylamine, 4-Anilinoaniline, 4-Anilinophenylamine, 4ADPA, Azoic Diazo Component 22, C.I. 76085, C.I. Developer 15, C.I. Oxidation Base 2, Diphenyl Black, Luxan Black R, N,4'-Bianiline, N-(4-Aminophenyl)aniline, N-Phenyl-1,4-benzenediamine, N-Phenyl-1,4-phenylenediamine, N-Phenyl-p-aminoaniline, N-Phenyl-p-phenylenediamine, NSC 3401, NSC 37074, Peltol BR, Peltol BR II, S 789, Semidin, Semidine, p-(Phenylamino)aniline, p-Aminodiphenylamine, p-Anilinoaniline, p-Semidine |
| InChI formel | InChI=1S/C12H12N2/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9,14H,13H2 |
| LEDER | Nc1ccc(Nc2ccccc2)cc1 |
| IUPAC-namn | 4-N-fenylbensen-1,4-diamin |
| Molekylvikt (g/mol) | 184.24 |
| Formel vikt | 184.1 |