Läs mer
N-Phenylmethyl-7-methyluric Acid, TRC
Produktkod. 30416728
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30416728 10 mg
30409513 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30416728 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP33577510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Phenylmethyl-7-methyluric Acid
CAS 72816-95-6
Molekylformel C13H12N4O3
Kvantitet 10 mg
Synonym 7,9-Dihydro-7-methyl-3-(phenylmethyl)-1H-purine-2,6,8(3H)-trione
InChI formel InChI=1S/C13H12N4O3/c1-16-9-10(14-12(16)19)17(13(20)15-11(9)18)7-8-5-3-2-4-6-8/h2-6H,7H2,1H3,(H,14,19)(H,15,18,20)
LEDER CN1C2=C(NC1=O)N(C(=O)NC2=O)CC3=CC=CC=C3
IUPAC-namn 3-benzyl-7-methyl-9H-purine-2,6,8-trione
Molekylvikt (g/mol) 272.26
Formel vikt 272.0909