Läs mer
N-Piperacillinyl Ampicillin, TRC
Produktkod. 30418195
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30418195 25 mg
1 options
Produktkod. 30418195 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP47996025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Piperacillinylampicillin
CAS 65772-67-0
Molekylformel C39 H44 N8 O10 S2
Kvantitet 25 mg
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[[[[[6-[[[[(4-ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl]carbonyl]amino]phenylacetyl]amino]-3,3-dimethyl-7-oxo-','[2S-[2α[S*(2R*,5S*,6S*)],5α,6β(S*)]]- (9CI), [2S-[2α[S*(2R*,5S*,6S*)],5α,6β(S*)]]-6-[[[[[6-[[[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl]carbonyl]amino]phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, N-Piperacillinyl Ampicillin, (2S,5R,6R)-6-[[(2R)-2-[[[(2S,5R,6R)-6-[[(2R)-2-[[(4-Ethyl-2,3-dioxopiperazin-1-yl)carbonyl]amino]- 2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl]carbonyl]amino]- 2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid, Piperacillinylampicillin
InChI formel InChI=1S/C39H44N8O10S2/c1-6-44-17-18-45(33(54)32(44)53)37(57)43-22(20-15-11-8-12-16-20)28(49)42-23-30(51)46-25(38(2,3)58-34(23)46)29(50)40-21(19-13-9-7-10-14-19)27(48)41-24-31(52)47-26(36(55)56)39(4,5)59-35(24)47/h7-16,21-26,34-35H,6,17-18H2,1-5H3,(H,40,50)(H,41,48)(H,42,49)(H,43,57)(H,55,56)/t21-,22-,23-,24-,25+,26+,34-,35-/m1/s1
LEDER CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(=O)N[C@@H](C(=O)N[C@H]4[C@H]5SC(C)(C)[C@@H](N5C4=O)C(=O)O)c6ccccc6)c7ccccc7)C(=O)C1=O
IUPAC-namn (2S,5R,6R)-6-[[(2R)-2-[[(2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molekylvikt (g/mol) 848.94
Formel vikt 848.2622