Läs mer
N-Pivaloylaniline, TRC
Produktkod. 30421363
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30421363 1 g
1 options
Produktkod. 30421363 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP5506651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Pivaloylaniline
CAS 6625-74-7
Molekylformel C11 H15 N O
Kvantitet 1 g
Synonym Propanamide','2,2-dimethyl-N-phenyl-, Pivalanilide (6CI,7CI,8CI), 2,2-Dimethyl-N-phenylpropanamide, N-Phenylpivalamide, N-Phenyltrimethylacetamide, N-Pivaloylaniline, NSC 58513, NSC 9043, PMSA 955, Pivaloylaminobenzene
InChI formel InChI=1S/C11H15NO/c1-11(2,3)10(13)12-9-7-5-4-6-8-9/h4-8H,1-3H3,(H,12,13)
LEDER CC(C)(C)C(=O)Nc1ccccc1
IUPAC-namn 2,2-dimethyl-N-phenylpropanamide
Molekylvikt (g/mol) 177.24
Formel vikt 177.1154